Would you like to log in again?
Difco™ Nutrient Broth, BD
Supplier: BD Diagnostics
Difco™
234000 234000 231000 232000 233000 233000 231000 232000
90002-660EA 336.16 USD InStock
90002-660 CA90002-592 90002-592 CA90002-108 90002-108 CA90002-622 90002-622 CA90002-660
Difco™ Nutrient Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Nutrient Broth is used for the cultivation of many species of nonfastidious microorganisms.
  • Beef extract provides carbohydrates, vitamins, nitrogen, and salts
  • Peptones provide a source of nitrogen
Nutrient Broth is one of several nonselective media useful in routine cultivation of microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty