Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
Antibiotics and Antimycotics, BD Diagnostics
Supplier: BD Diagnostics
231651 231289 231593 231213 231664 231633 231628 231562 231290 231597 231657 231577 230793 231268 231562 230705 231275 231552 230733 231591 231635 231641 231591 231597 231613 231682 231290 231264 231606 231658 231593 231040 231278 231042 231278 231552 231041 231274 231268 231590 231658 231271 230749 231629 231607 231122 231041 231635 231286 231275 231664 231641 231613 230725 231607 231263 231629 231660 231633 231264 231555 231286 231682 231660 231289 231263 231271 231274
90002-342EA 63.4 USD InStock
90002-342 90002-062 90002-382 CA90002-486L CA90002-428 CA90002-504 CA90002-228 90002-334 90002-214 CA90002-342 90002-396 90002-474 90002-078 CA90002-240 90002-230 CA90002-480 90002-074 90002-072 CA90002-398L CA90002-334L 90002-428 CA90002-118 90002-426 CA90002-436 CA90002-238 90002-228 90002-504 CA90002-236 CA90002-432 CA90002-234 90002-464 90002-068 CA90002-250 90002-240 CA90002-370 90002-120 90002-480 CA90002-230L CA90002-476L CA90002-408 90002-118 CA90002-446 CA90002-204 CA90002-248 90002-116 90002-436 CA90002-246 90002-238 CA90002-244 90002-236 CA90002-364 90002-398 90002-234 90002-476 90002-432 90002-374 CA90002-340 CA90002-382 90002-370 90002-250 90002-408 90002-248 90002-446 90002-246 90002-368 90002-244 90002-486 CA90002-374
Antibiotics and Antimycotics, BD Diagnostics
Microbiology Test Systems Antibiotic Tests
Ideal for the qualitative susceptibility testing of mycobacteria and related organisms.
  • Reliable
  • Convenient
  • Rapid method
This collection of media products is used for the evaluation of susceptibility of microorganisms to anti-microbial agents.
Packaging: Each test includes 50 susceptibility disks per cartridge.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty