Would you like to log in again?
Difco™ Novobiocin Antimicrobic Supplement, BD
Supplier: BD Life Sciences
Difco™
231971
90002-590EA 495.03 USD InStock
90002-590
Difco™ Novobiocin Antimicrobic Supplement, BD
Media for Microbiology Prepared Media for Microbiology
Novobiocin Antimicrobic Supplement is used with EC Medium Modified, Rappaport-Vassiliadis Medium Semisolid Modification and Brilliant Green Agar
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty