Would you like to log in again?
Difco™ Skim Milk, BD Life Sciences
Supplier: BD Life Sciences
232100 232100
90002-594EA 86.63 USD InStock
90002-594 CA90002-594
Difco™ Skim Milk, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Differentiation of organisms based on coagulation and proteolysis of casein
Proteolytic bacteria use the enzyme caseinase to hydrolyze casein and form soluble nitrogenous compounds displayed as a clear zone around colonies. This clearing is much more pronounced on agar containing milk if the bacteria are able to produce acid from fermentable carbohydrates in the medium. Moreover, the hydrolysis of casein is often used to evaluate proteolytic activity of psychrotrophic microorganisms of significance to food, water, and dairy industries.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty