Would you like to log in again?
Difco™ Niacin Assay Medium, BD
Supplier: BD Life Sciences
Difco™
232210 232210
90002-600EA 101.83 USD InStock
CA90002-600 90002-600
Difco™ Niacin Assay Medium, BD
Media for Microbiology Prepared Media for Microbiology
Niacin Assay Medium is used for determining niacin concentration by the microbiological assay technique.
  • Dehydrated medium free from nicotinic acid
  • Addition of nicotinic acid stimulates growth response
  • Very light amber appearance when prepared
Niacin Assay Medium is a dehydrated medium free from nicotinic acid and its analogs but containing all other nutrients and vitamins essential for the cultivation of L. plantarum ATCC™ 8014.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty