Would you like to log in again?
Rosolic Acid, BD
Supplier: BD Diagnostics
232281 232281
90002-602PK 507.28 USD InStock
CA90002-602 90002-602
Rosolic Acid, BD
Media for Microbiology Dehydrated Media for Microbiology
Rosolic Acid is used with m FC Agar and m FC Broth Base in cultivating and enumerating fecal coliforms by the membrane filter technique at elevated temperatures.
  • Practically insoluble in water
  • Freely soluble in alcohol
  • Can be used as a pH indicator
Soluble in strong acids to form yellow solution, or in aqueous alkalis to form carmine red solutions. Because of its behavior in changing color due to the presence of an acidic or alkaline solution, rosolic acid is used as a pH indicator with pH transition range of 5.0-6.8.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty