Would you like to log in again?
Riboflavin Assay Medium, BD
Supplier: BD Diagnostics
232510 232510
90002-608EA 197.08 USD InStock
CA90002-608 90002-608
Riboflavin Assay Medium, BD
Media for Microbiology Dehydrated Media for Microbiology
Riboflavin Assay Medium is used for determining riboflavin concentration by the microbiological assay technique.
  • Dehydrated appearance: Beige, free-flowing
  • Free of riboflavin but contains all other nutrients and vitamins essential for the growth of Lactobacillus rhamnosus
The addition of riboflavin in specified increasming concentrations gives a growth response that can be measured turbidimetrically or titrimetrically.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty