Would you like to log in again?
Difco™ Antimicrobic Vial A Chlortetracycline, BD Diagnostics
Supplier: BD
233331 233331
90002-632PK 340.33 USD InStock
CA90002-632 90002-632
Difco™ Antimicrobic Vial A Chlortetracycline, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Difco™ Antimicrobic Vial A, containing chlortetracycline, is used in preparing selective microbiological culture media.
  • Contains sterile, desiccated chlortetracycline
  • Applicable for use in various media requiring this antibiotic
  • Preferred in Cooke Rose Bengal Agar due to increased stability
Antimicrobic Vial A was originally used in preparing DTM Agar described by Taplin, Azias, Rebell, and Blank for the isolation of dermatophytes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty