Would you like to log in again?
Difco™ Antimicrobic Vial K Kanamycin, BD Diagnostics
Supplier: BD
233391 233391
90002-634PK 661.35 USD InStock
CA90002-634 90002-634
Difco™ Antimicrobic Vial K Kanamycin, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Antimicrobic Vial K Kanamycin is effective as a bacteriocidal agent by inhibiting ribosomal translocation and eliciting miscoding.
  • Inhibits growth of gram-negative bacteria
  • Used to isolate resistant bacteria
Difco™ Antimicrobic Vial K, containing kanamycin, is used to supplement SFP Agar Base containing Egg Yolk Enrichment 50%.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty