Would you like to log in again?
Difco™ Supplement VX with Reconstituting Fluid, BD Biosciences
Supplier: BD Biosciences
233542 233541 233542 233541
90002-650PK 706.76 USD InStock
90002-650 CA90002-652 CA90002-650 90002-652
Difco™ Supplement VX with Reconstituting Fluid, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Supplement VX with Reconstituting Fluid is used for supplementing media to culture fastidious microorganisms, particularly Neisseria gonorrhoeae and Haemophilus influenzae.
  • Lyophilized concentrate
  • Contains vitamins, amino acids, coenzymes, and dextrose
This supplement is recommended for enriching GC Agar media, Proteose No. 3 Agar, and the selective agars for the isolation of pathogenic Neisseria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty