Would you like to log in again?
Difco™ Tinsdale Base, BD Biosciences
Supplier: BD Biosciences
278610 234210 278610 234210
90003-990EA 693.04 USD InStock
90003-990 CA90002-668 90002-668 CA90003-990
Difco™ Tinsdale Base, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Tinsdale Agar base is used with Tinsdale Enrichment, Desiccated in isolating and differentiating Corynebacterium diphtheriae.
  • Peptone provides nitrogen, vitamins, carbon, and amino acids
  • Sodium chloride maintains the osmotic balance of the medium
  • Agar is the solidifying agent
Tinsdale Enrichment, Dessicated contains bovine serum and horse serum, which provide essential growth factors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty