Would you like to log in again?
Difco™ Tryptic Soy Agar, BD Biosciences
Supplier: BD Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
236930 236920 236950 236920 236950 236940
90002-706EA 169.37 USD InStock
CA90002-706 CA90002-704 90002-706 CA90002-700 90002-702 90002-700
Difco™ Tryptic Soy Agar, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Tryptic Soy Agar is used for the isolation and cultivation of nonfastidious and fastidious microorganisms.
  • Casein and soy peptones promote general growth
  • Sodium chloride maintains the osmotic equilibrium
Tryptic Soy Agar is not the medium of choice for anaerobes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty