Would you like to log in again?
Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD Diagnostics
237400 237500 237400 237500 237200 237200
90003-032EA 781.56 USD InStock
CA90003-040 90003-032 90003-040 90003-038 CA90003-038 CA90003-032
Bacto™ Brain Heart Infusion, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Brain Heart Infusion (BHI) Broth is a general-purpose liquid medium for the growth of a wide variety of bacterial and fungal species.
  • Nutrients come from peptones and infusions of brain and heart
  • Supports the growth of fastidious and nonfastidious microorganisms
BHI Broth is used for the cultivation of a wide variety of microorganisms, including bacteria, yeasts, and molds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty