Would you like to log in again?
Difco™ Sabouraud Dextrose Broth, BD
Supplier: BD Diagnostics
238230 238230 238210 238210
90003-096EA 141.94 USD InStock
90003-096 CA90003-096 90003-092 CA90003-092
Difco™ Sabouraud Dextrose Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Dextrose Broth is used for the cultivation of yeasts, molds, and aciduric microorganisms.
  • Peptone provides nitrogen, vitamins, minerals, amino acids, and growth factors
  • Low pH promotes the growth of fungi
  • Dehydrated appearance: Cream to yellow, free-flowing
The addition of antimicrobics further increases the selectivity of the medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty