Would you like to log in again?
BD BBL™ Dubos Broth Base, BD
Supplier: BD Biosciences
238510
90003-100EA 399.32 USD InStock
90003-100
BD BBL™ Dubos Broth Base, BD
Media for Microbiology Dehydrated Media for Microbiology
Dubos broth base is used with dubos medium albumin for rapidly cultivating pure cultures of Mycobacterium tuberculosis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty