Would you like to log in again?
Difco™ Azide Dextrose Broth, BD
Supplier: BD Life Sciences
238710 238710
90003-102EA 280.95 USD InStock
CA90003-102 90003-102
Difco™ Azide Dextrose Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Azide Dextrose Broth is used for cultivating streptococci in water and wastewater.
  • Beef extract and peptones provide essential nutrients
  • Dextrose is a fermentable carbohydrate
  • Sodium chloride maintains osmotic equilibrium
  • Sodium azide inhibits cytochrome oxidase in gram-negative bacteria
Azide Dextrose Broth is specified for use in the presumptive test of water and wastewater for fecal streptococci by the Multiple-Tube Technique.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty