Would you like to log in again?
Difco™ Malonate Broth, BD Diagnostics
Supplier: BD Diagnostics
Difco™
239520 239520
90003-110EA 517.76 USD InStock
90003-110 CA90003-110
Difco™ Malonate Broth, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Malonate Broth is used for differentiating Enterobacter from Escherichia based on malonate utilization.
  • Contains ammonium sulfate as a sole source of nitrogen
  • Dipotassium phosphate and monopotassium phosphate provide buffering capability
  • Sodium chloride maintains osmotic equilibrium
  • Bromthymol blue is a pH indicator
Malonate Broth is described for differentiating Enterobacteriaceae in food and dairy products, and is suitable for organisms that would otherwise fail on the unsupplemented Leifson medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty