Would you like to log in again?
Difco™ Brain Heart Infusion Agar, BD Life Sciences
Supplier: BD Life Sciences
241810 241820 241830 241830 241810
90003-262EA 363.67 USD InStock
CA90003-260 90003-262 CA90003-262 90003-258 CA90003-258
Difco™ Brain Heart Infusion Agar, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Brain Heart Infusion Agar is a general-purpose culture medium which supports the growth of a wide variety of bacterial and fungal species including many types of pathogens, such as streptococci and pneumococci.
  • Brain heart infusion, peptones, and dextrose provide nutrients
  • Dextrose is the carbohydrate source for fermentation
  • Disodium phosphate acts as a buffering agent
Brain Heart Infusion (BHI) Agar is a general-purpose medium suitable for the cultivation of a wide variety of organism types, including bacteria, yeasts, and molds. With the addition of 10% sheep blood, it is used for the isolation and cultivation of a wide variety of fungal species, including systemic fungi, from clinical and nonclinical sources.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty