Would you like to log in again?
Difco™ Biotin Assay Medium, BD
Supplier: BD Life Sciences
241910 241910
90003-264EA 130.66 USD InStock
90003-264 CA90003-264
Difco™ Biotin Assay Medium, BD
Media for Microbiology Dehydrated Media for Microbiology
Biotin Assay Medium is used for determining biotin concentration by the microbiological assay technique.
  • Biotin-free dehydrated medium
  • Addition of biotin gives a growth response
  • Dehydrated appearance: Light beige, homogeneous
Biotin Assay Medium may be used for both turbidimetric and titrimetric analysis. The most effective assay range, using Biotin Assay Medium, has been found to be between 0.1 ng and 1 ng biotin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty