Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lowenstein Medium, BD Diagnostics
Supplier: BD Diagnostics
244420 244410 244410 244420
90003-340EA 855.14 USD InStock
90003-340 90003-342 CA90003-340 CA90003-342
Lowenstein Medium, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Isolation and differentiation of mycobacteria.
  • Use with Glycerol
  • Prepare a variety of coagulated egg media
Prepared, tubed and ampuled media available. Use with Glycerol to prepare a variety of coagulated egg media for isolation and differentiation of mycobacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty