Would you like to log in again?
Pseudomonas Agar F, BD
Supplier: BD Diagnostics
244820 244910 244910 244820
90003-352EA 411.18 USD InStock
90003-352 CA90003-352 90003-354 CA90003-354
Pseudomonas Agar F, BD
Media for Microbiology Dehydrated Media for Microbiology
Pseudomonas Agar F, also known as Flo Agar, is used for the enhancement of fluorescin production and Pseudomonas Agar P, also known as Tech Agar, is used for the enhancement of pyocyanin production by Pseudomonas.
  • 1:1 ratio of casein to meat peptone promotes fluorescin production
  • Magnesium sulfate provides ions for fluorescin production
  • Enzymatic digest of gelatin provides nitrogen and amino acids
  • Magnesium, potassium, and sulfate ions promote pyocyanin production
Both media contain glycerol, which acts as a source of energy and enhances pigment production.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty