Would you like to log in again?
BBL™ MGIT™ Mycobacteria Growth Indicator, BD Diagnostics
Supplier: BD Diagnostics
245116 245113 245114 245111 245114 245113 245116 245111
90003-371PK 2117.81 USD InStock
CA-90003-368 90003-371 90003-372 90003-374 CA-90003-374 90003-368 CA-90003-370 CA-90003-372
BBL™ MGIT™ Mycobacteria Growth Indicator, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
The BBL™ Mycobacteria Growth Indicator Tube is a rapid, easy-to-use system with high accuracy for the detection of mycobacteria directly from clinical specimens.
  • Growth in as little as four to six days
  • Unmistakable results
  • Outstanding performance
  • Fits perfectly into your laboratory work flow
The BBL™ MGIT™ Mycobacteria Growth Indicator Tube supplemented with BBL™ MGIT™ OADC enrichment and BBL™ MGIT™ PANTA™ antibiotic mixture, when appropriate, is intended for the detection and recovery of mycobacteria.
Acceptable specimen types are digested and decontaminated clinical specimens (except urine) and sterile body fluids (except blood).
BBL™ MGIT™ tubes also fit into the standard procedures for decontamination (NaCl-NaOH) and disposal of waste. It is ideal for any size laboratory, especially those performing a high volume of mycobacteria tests.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty