Would you like to log in again?
Difco™ Antibiotic Medium, BD Life Sciences
Supplier: BD Life Sciences
Difco™
227710 292940 259310 224410 224410 266710 227020 226340 227020 226340 266910 266710 227710 243100 246310 259310 246310 246210 246210 266910 243100
90003-548EA 255.6 USD InStock
90003-548 CA90003-548 90003-786 CA90003-786 90001-890 CA90001-890 CA90003-392 90003-790 CA90003-292 CA90003-790 90001-788 90001-920 90003-390 90003-292 CA90003-390 CA90001-788 CA90001-920 90003-392 90001-938 CA90001-938 90004-466
Difco™ Antibiotic Medium, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Antibiotic assay media are used for determining antibiotic potency by the microbiological assay technique.
  • Potency is tested by demonstrating its inhibitory effect
Antibiotic medium 1 and antibiotic medium 4 are used in a cylinder plate method for detecting penicillin in nonfat dry milk.
Antibiotic medium 12 is prepared with the grove and randall formula. It is used for the assay of amphotericin B. Recommended for use in preparing test plates for the cylinder plate assay of nystatin and anisomycin.
Antibiotic medium 19 corresponds to the use described in outline of details for official microbiological assays of antibiotics
Cylinder plate assays are used for commercial preparations of antibiotics, as well as in the quantitative determination of antibioitics in body fluids, animal feeds, and other materials.
Certifications: Antibiotic media are prepared according to the specifications of the USP, European Pharmacopeia, and AOAC International.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty