Would you like to log in again?
Difco™ WL Nutrient Broth, BD
Supplier: BD Life Sciences
247110 247110
90003-400EA 212.93 USD InStock
CA90003-400 90003-400
Difco™ WL Nutrient Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Difco™ WL Nutrient Broth is used for the cultivation of yeasts, molds and bacteria encountered in brewing and industrial fermentation processes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty