Would you like to log in again?
Rogosa SL Agar, BD
Supplier: BD Diagnostics
248020 248020
90003-412EA 322.18 USD InStock
90003-412 CA90003-412
Rogosa SL Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Rogosa SL Agar is used for cultivating oral, vaginal, and fecal lactobacilli.
  • Peptone provides carbon and nitrogen
  • Carbohydrate sources provide extra carbon
  • Polysorbate 80 acts as a surfactant
  • Agar is the solidifying agent
  • Low pH of the medium suppresses other bacterial flora
These media are used for the isolation, enumeration, and identification of lactobacilli in oral bacteriology, feces, vaginal specimens, and foodstuffs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty