Would you like to log in again?
Difco™ Tetrathionate (TT) Broth Base, Hajna, BD Biosciences
Supplier: BD Biosciences
249120 249120
90003-424EA 191.09 USD InStock
90003-424 CA90003-424
Difco™ Tetrathionate (TT) Broth Base, Hajna, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
TT Broth Base, Hajna is used for enriching Salmonella from food and dairy products prior to isolation procedures.
  • Peptone provides nitrogen and amino acids
  • Yeast extract supplies growth factors and vitamins
  • Dextrose and mannitol are fermentable carbohydrates
  • Calcium carbonate is a neutralizer that absorbs toxic metabolites
Organisms containing the enzyme tetrathionate reductase will proliferate in this medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty