Would you like to log in again?
Bacto™ Todd Hewitt Broth, BD Diagnostics
Supplier: BD Diagnostics
249240 249240 249220
90003-428EA 3167.63 USD InStock
90003-428 90003-430 CA90003-430
Bacto™ Todd Hewitt Broth, BD Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Todd Hewitt Broth is a general-purpose medium, which primarily is used for the cultivation of beta-hemolytic strepto-cocci, especially for serological studies.
  • Contains peptones, dextrose, and salts for promoting general growth
  • Dehydrated apperarance: Light beige, free-flowing
Selective enrichment broths include the advantages of both enrichment and selection by providing conditions conducive to the growth of group B streptococci while inhibiting the growth of contaminants.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty