Would you like to log in again?
Difco™ Cystine Tryptic Agar, BD Life Sciences
Supplier: BD Life Sciences
Difco™
252310 252310
90003-460EA 400.32 USD InStock
90003-460 CA90003-460
Difco™ Cystine Tryptic Agar, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Cystine tryptic agar is used with added carbohydrates in differentiating microorganisms based on fermentation reactions and motility.
This agar is used for the maintenance of microorganisms, as well as for the detection of bacterial motility and, with added carbohydrate, for fermentation reactions of fastidious microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty