Would you like to log in again?
Difco™ B12 Assay Medium, BD Life Sciences
Supplier: BD Life Sciences
254110 245710 245710 254110
90003-384EA 168.96 USD InStock
90003-384 90003-484 CA90003-484 CA90003-384
Difco™ B12 Assay Medium, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
B12 Assay Medium is used for determining vitamin B12 concentration by the microbiological assay technique.
  • Dehydrated appearance: Very light to light beige, homogeneous
  • 4.25% single strength solution
  • Solution is light amber, clear, may have a slight precipitate
B12 Assay Medium is used in the microbiological assay of vitamin B12 according to the procedures of the Vitamin B12 Activity Assay in the USP1 and the Cobalamin (Vitamin B12 Activity) Assay in the Official Methods of Analysis of AOAC International (AOAC).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty