Would you like to log in again?
Difco™ PPLO Media (Mycoplasma Media), BD
Supplier: BD Diagnostics
211456 241210 241210 255420 255420 211456
90003-504EA 260.73 USD InStock
90003-504 CA90003-504 90003-248 CA90003-248 CA90000-198 90000-198
Difco™ PPLO Media (Mycoplasma Media), BD
Media for Microbiology Dehydrated Media for Microbiology
PPLO (Mycoplasma) agars and broths, when supplemented with nutritive enrichments, are used for isolating and cultivating Mycoplasma.
  • Meat digests, peptones, beef extract, and yeast extract provide nutrients
  • Sodium chloride maintains osmotic equilibrium
Agar, the solidifying agent, is used in PPLO (Mycoplasma) Agar at a concentration slightly reduced from usual to ensure formation of the largest possible colonies because the organisms grow into the agar with only slight surface growth.
Mycoplasma Supplement contains fresh yeast extract and horse serum. Yeast extract provides the preformed nucleic acid precursors that are required by Mycoplasma species. Horse serum supplies cholesterol, a growth stimulant.
Mycoplasma Enrichment without Penicillin is a selective enrichment containing the inhibitor thallium acetate, to which a penicillin of choice can be added at the time of use to make it selective against gram-positive and gram-negative bacteria.
Mycoplasma Broth Base (Frey) is used for the cultivation of avian mycoplasmas.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty