Would you like to log in again?
Bacto™ TC Yeastolate, Life Technologies
Supplier: Life Technologies
255772 255771 255772 255771
90003-508EA 3250 USD InStock
90003-508 CA90003-508 CA90003-510 90003-510
Bacto™ TC Yeastolate, Life Technologies
Media for Microbiology Dehydrated Media for Microbiology
Bacto™ TC Yeastolate is an animal-free hydrolysate from the water-soluble portion of autolyzed yeast
  • Animal-free
  • Water-soluble portion of autolyzed yeast
  • Ultra-filtered
  • For CHO-based production of antibodies
  • Provides optimal titer performance in mammalian cell-based processes
This supplement contains a mixture of peptides, amino acids, carbohydrates (simple and complex), neucleosides, and vitamins. Derived from the same yeast strain as Difco™ TC Yeastolate, Ultra-Filtered, this product has a slightly different nutriential profile than the ultrafiltered product because it was not ultrafiltered. Bacto™ TC Yeastolate should be considered a suitable alternative to Difco™ TC Yeastolate, Ultra-Filtered, for supplementing a variety of CHO-based applications. Customers have used this supplement for CHO-based production of biotherapeutic monoclonal antibodies used in oncological and rheumatological therapeutics and recombinant proteins. Additionally, customers have used this supplement in combination with a soy supplement, such as Difco™ Select Phytone UF, to provide an optimal titer performance in mammalian cell based processes by providing additional vitamins and carbohydrates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty