Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BD BBL™ Diagnostic Reagent and Stain Droppers, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™
261185 261198 261182 261188 261204 261181 261192 261191 261183 261190 261201 261191 261195 261195 261203 261193 261196 261197 261204 261203 261193 261189 261185 261198 261181 261196 261183 261197 261189 261182 261187 261192 261194 261188 261187 261194 261190 261201
95025-232PK 158.68 USD InStock
95025-232 CA90003-678 CA90003-658 90003-688 CA90003-674 90003-666 90003-668 CA90003-676 90003-662 90003-684 CA90003-692 CA90003-670 CA90003-672 90003-664 90003-686 90003-690 90003-692 90003-670 CA90003-680 90003-658 CA90003-668 CA90003-664 90003-676 CA90003-686 90003-678 CA90010-088 CA90003-688 CA90003-666 CA90003-660 CA90003-682 90003-672 90003-674 CA90003-684 CA90003-662 90003-681 90003-682 90003-660 CA90003-690
BD BBL™ Diagnostic Reagent and Stain Droppers, BD Diagnostics
Stains Microbiology Stains
Reagents are hermetically sealed in an ampule to protect from chemical instability
They are used by holding the dropper upright and squeezing it to crush the ampule inside. The dropper can then be tilted for convenient, drop-by-drop dispensing. Each ampule contains a 1-day supply of premixed reagent and has a shelf life of one to three years without refrigeration. Flat bottoms allow the droppers to stand upright on the benchtop.
Ordering information: Each reagent comes in a box of 50 ampules.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty