Would you like to log in again?
Modified Letheen Broth, BD Biosciences
Supplier: BD Biosciences
263010 263010
90003-730EA 284.67 USD InStock
CA90003-730 90003-730
Modified Letheen Broth, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Dehydrated Culture Media for microbiological evaluation of cosmetics
Inactivates preservative agents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty