Would you like to log in again?
Difco™ TCBS (Thiosulfate-Citrate-Bile-Sucrose) Agar, BD Biosciences
Supplier: BD Biosciences
265020 265020
90003-770EA 283.18 USD InStock
90003-770 CA90003-770
Difco™ TCBS (Thiosulfate-Citrate-Bile-Sucrose) Agar, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
TCBS Agar is used for the selective isolation of cholera vibrios and Vibrio parahaemolyticus from a variety of clinical and nonclinical specimens.
  • Oxgall inhibits the growth of Gram-positive bacteria
  • Sodium thiosulfate serves as a sulfur source
  • Sodium thiosulfate detects hydrogen sulfide production
  • Sucrose is included as a source of carbohydrates
  • Alkaline pH enhances the recovery of V. cholerae
Thymol blue and bromthymol blue are included as indicators of pH changes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty