Would you like to log in again?
Difco™ Demi-Fraser Broth Base, Dehydrated Culture Medium, BD Diagnostics
Supplier: BD Diagnostics
Difco™
265320 265310 265320
90003-776EA 216.06 USD InStock
90003-776 CA90003-776 90003-774
Difco™ Demi-Fraser Broth Base, Dehydrated Culture Medium, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Difco™ Demi-Fraser Broth Base is for use with Fraser Broth Supplement in selectively and differentially enriching Listeria in microbiology applications.
  • Reduced Acriflavine and Nalidixic Acid levels
  • For Listeria enrichment
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty