Would you like to log in again?
Difco™ m FC Agar, BD Diagnostics
Supplier: BD Diagnostics
Difco™
267710 267720 267710 267720
90003-794EA 130.6 USD InStock
CA90003-796 90003-794 CA90003-794 90003-796
Difco™ m FC Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Use with Rosolic Acid
For detection and enumeration of fecal coliforms by the membrane filter technique at elevated temperatures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty