Would you like to log in again?
Difco™ Fluid Selenite-Cystine Broth, BD
Supplier: BD Life Sciences
268740 268740
90003-808EA 541.06 USD InStock
90003-808 CA90003-808
Difco™ Fluid Selenite-Cystine Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Fluid Selenite-Cystine Broth is used as an enrichment medium for isolation of Salmonella species from foods, dairy products, materials of sanitary importance, and clinical specimens.
  • Selective enrichment for the cultivation of Salmonella species
  • Must not be autoclaved
  • Casein peptone provides essential nitrogenous and carbon compounds
Selenite-Cystine Broth is used as an enrichment medium for the isolation of Salmonella from feces, urine, water, foods, and other materials of sanitary importance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty