Would you like to log in again?
Difco™ Bismuth Sulfite Agar, BD
Supplier: BD Life Sciences
273300 273300
90003-904EA 287.74 USD InStock
90003-904 CA90003-904
Difco™ Bismuth Sulfite Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Bismuth Sulfite Agar is a highly selective medium used for isolating Salmonella species, particularly Salmonella Typhi, from food and clinical specimens.
  • Modification of the Wilson and Blair formula
  • Beef extract and peptone provide nitrogen, vitamins, and minerals
  • Dextrose is an energy source
  • Disodium phosphate is a buffering agent
  • Brilliant green and bismuth sulfite inhibit gram-positive bacteria
  • Ferrous sulfate is included for detection of H2S production
Bismuth Sulfite Agar is used for the isolation of S. Typhi and other Salmonella from food, feces, urine, sewage, and other infectious materials. The typhoid organism grows luxuriantly on the medium, forming characteristic black colonies, while gram-positive bacteria and members of the coliform group are inhibited.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty