Would you like to log in again?
Macro-Vue™ RPR Circle Card Test Kits, BD Diagnostics
Supplier: BD Diagnostics
Macro-Vue™
275005 275539 275005 274449 275539 274449 275239
90003-956PK 902.21 USD InStock
CA90003-924 90003-956 90003-924 CA90003-960 90003-942 CA90003-942L 90003-960
Macro-Vue™ RPR Circle Card Test Kits, BD Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
These rapid plasma reagin (RPR) test kits are developed for use in large-scale testing in public health and clinical laboratories
Kits use mechanical rotation, antigen suspension, and specially prepared, plastic-coated test cards for the serologic detection of syphilis. The recommended speed for rotation is 100rpm. The rotator should circumscribe a circle approximately 2cm in diameter in the horizontal plane. Refrigeration is recommended for storing the antigen suspension ampules. The disposable BD Dispenstirs™ devices allow the transfer of specimen to the card surface. An antigen dispensing bottle with a needle serves for accurate drop delivery of antigen. Spot cards feature circle spots 18mm in diameter.
Kit contents:
3 mL Antigen
Ampules
10-Spot Cards
Capillaries, 0.05 mL
Antigen Dispensing Bottle Stirrers
20-Gauge Dispensing Needle
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty