Would you like to log in again?
Difco™ Tellurite Enrichment Medium, BD Life Sciences
Supplier: BD Life Sciences
277910 277910
90003-984PK 647.07 USD InStock
90003-984 CA90003-984
Difco™ Tellurite Enrichment Medium, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Used with Baird-Parker Agar Base for the isolation of S
aureus. The enrichment consists of 30% egg yolk suspension with 0.15% potassium tellurite.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty