Would you like to log in again?
Difco™ Columbia Blood Agar, BD
Supplier: BD Life Sciences
279030 279230 279020 279220 279030 279220
90004-008EA 783.09 USD InStock
CA90004-008 90004-008 90004-010 90004-002 90004-000 CA90004-002
Difco™ Columbia Blood Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Columbia Blood Agar is an infusion-free basal medium to use with or without blood for the cultivation of fastidious microorganisms.
  • Used with blood in isolating and cultivating fastidious microorganisms
  • Used in qualitative procedures
  • For isolating and cultivating Haemophilus species from clinical specimens
Columbia Blood Agar is utilized as the base for media containing blood and for selective media formulations in which various combinations of antimicrobial agents are used as additives.
Columbia Blood Agar Base EH (Enhanced Hydrolysis) is used with blood in isolating and cultivating fastidious microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty