Would you like to log in again?
Difco™ Leptospira Enrichment EMJH, BD Diagnostics
Supplier: BD Diagnostics
279510 279510 279410
90004-014EA 301.91 USD InStock
90004-014 CA90004-016 90004-016
Difco™ Leptospira Enrichment EMJH, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Leptospira Enrichment is a filter-sterilized chemical enrichment supplement recommended for cultivation of Leptospira species.
  • May be lyophilized
  • 10 mL per vial
  • Must be refrigerated upon arrival
Leptospira Enrichment can come lyophilized and is sterilized for ready use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty