Would you like to log in again?
Difco™ MacConkey Agar Base, BD Diagnostics
Supplier: BD Diagnostics
Difco™
281810 281810
90004-036EA 330.13 USD InStock
90004-036 CA90004-036
Difco™ MacConkey Agar Base, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
MacConkey Agar Base is a solid medium recommended for use in qualitative procedures for the cultivation of gram-negative bacilli.
  • Peptones provide nitrogenous nutrients and amino acids
  • Sodium chloride maintains osmotic equilibrium
  • Crystal violet and bile salts inhibit most gram-positive organisms
MacConkey Agar Base is used with added carbohydrate to differentiate enteric gram-negative bacilli based on fermentation reactions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty