Would you like to log in again?
Difco™ Candida BCG Agar Base, BD
Supplier: BD Life Sciences
Difco™
283510
90004-044EA 348.05 USD InStock
90004-044
Difco™ Candida BCG Agar Base, BD
Media for Microbiology Dehydrated Media for Microbiology
Candida BCG Agar Base is for use with added neomycin for the primary isolation and identification of Candida species.
  • Differential and selective medium
  • Use for the primary isolation and identification of Candida species
This medium consists of peptone agar base supplemented with yeast extract and dextrose to provide the nutrients necessary to support growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty