Would you like to log in again?
BD Difco™ Mycoplasma Supplement, BD Diagnostics
Supplier: BD Diagnostics
283610
90004-046PK 2432.23 USD InStock
90004-046
BD Difco™ Mycoplasma Supplement, BD Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
These mycoplasma supplement are prepared from yeast extract and horse serum for the isolation and cultivation of Mycoplasma species
For use with PPLO agar and agar base, PPLO broth and broth base.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty