Would you like to log in again?
Lysine Medium, BD Diagnostics
Supplier: BD Diagnostics
211759 284920 211759 284920
90000-334EA 602.18 USD InStock
CA90000-334 90000-334 CA90004-052 90004-052
Lysine Medium, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Differential medium to detect organisms that decarboxylate or deaminate lysine and produce hydrogen sulfide.
Differentiation of bacteria based on ability to decarboxylate lysine.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty