Would you like to log in again?
Bacto™ Tryptic Soy Broth without Dextrose, Life Technologies
Supplier: Life Technologies
286220 286220
90004-068EA 207.13 USD InStock
90004-068 CA90004-068
Bacto™ Tryptic Soy Broth without Dextrose, Life Technologies
Media for Microbiology Additives and Supplements for Microbiology
Tryptic Soy Broth is a general-purpose liquid enrichment medium used in qualitative procedures for the sterility test and for the enrichment and cultivation of aerobic microorganisms that are not excessively fastidious.
  • Supports the growth of a wide variety of microorganisms
  • Enzymatic digests of casein and soybean meal provide amino acids and nitrogen
  • Lack of glucose means this mixture is low in carbohydrates
Growth in broth media is indicated by the presence of turbidity, specks, or flocculation in the medium while an uninoculated control remains clear and without turbidity after incubation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty