Would you like to log in again?
Difco™ Motility GI Medium, BD Life Sciences
Supplier: BD Life Sciences
Difco™
286910 286910
90004-070EA 255.62 USD InStock
CA90004-070 90004-070
Difco™ Motility GI Medium, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Motility GI Medium is used for detecting motility of microorganisms and for separating organisms in their motile phase.
  • Beef heart infusion, peptone, and gelatin provide nitrogen and vitamins
  • Agar is the solidifying agent
  • Motility evidenced by diffuse growth away from line of inoculation
Nonmotile organisms grow only along the line of inoculation; the presence of diffuse growth away from the line or spot of inoculation is evidence of motility.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty