Would you like to log in again?
Difco™ Decarboxylase Medium Base, BD Life Sciences
Supplier: BD Life Sciences
287220 287220
90004-072EA 400.79 USD InStock
90004-072 CA90004-072
Difco™ Decarboxylase Medium Base, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Decarboxylase media are used in the biochemical differentiation of gram-negative enteric bacilli based on the production of arginine dihydrolase and lysine and ornithine decarboxylase.
  • Peptones and beef or yeast extract provide nutrients
  • Pyridoxal is an enzyme co-factor
  • Dextrose is a fermentable carbohydrate
  • Bromcresol purple and cresol red are pH indicators
  • Amino acids are added to detect production of specific enzymes
Decarboxylase Medium Base, with added arginine, lysine, or ornithine is used for the same purpose.
Lysine Decarboxylase Broth is used for differentiating microorganisms based on lysine decarboxylation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty