Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
BD Hycheck™ Paddles for Monitoring Production Environments, BD Diagnostics
Supplier: BD Diagnostics
BD Hycheck™
290002 290006 290006 290004 290007 290005 290004 290003 290002 290007 290001 290003 290001 290005 290003
90004-108PK 90.94 USD InStock
CA89200-310 CA90004-108 CA90004-106 90004-108 90004-112 CA90004-100 90004-102 CA90004-110 90004-110 90004-100 CA90004-104 90004-106 CA90004-102 CA90004-112 90004-104
BD Hycheck™ Paddles for Monitoring Production Environments, BD Diagnostics
Microbiology Test Systems Microorganism Tests
BD HYcheck™ is a hygiene contact slide used to assess the microbiological contamination of surfaces or fluids
The double-sided slide features a hinged paddle that bends for easy sampling.
Ordering information: Store at 2 to 15°C (36 to 59°F). Multiple media combinations are available; contact your Avantor sales representative for details.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty